Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Talon®, Burette clamp
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR® Talon®
VWRU17683-258EA 5263 INR InStock
VWRU17683-258
VWR® Talon®, Burette clamp
Clamps General Purpose Clamps
Zinc. This double burette clamp holds any size burette from micro to 100 ml capacity. Simply compress the scissor-like mechanism, insert burette, and gently release to grip.
  • Numbers and graduation on burette remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release
This double burette clamp holds any size burette from micro to 100 ml capacity. Simply compress the scissor-like mechanism, insert burette, and gently release to grip.
  • Numbers and graduation on burette remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms