Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Culture tubes, round bottom
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
89000-496 89000-504
VWRU89000-496EA 0 INR InStock
VWRU89000-496 VWRU89000-504
VWR®, Culture tubes, round bottom
Tubes Culture Tubes
Borosilicate glass, Type I, Class A.
The culture tubes made with a fire-polished top rim and white ceramic enamel marking spot. The uniform wall thickness for maximum heat transfer and chemical resistance.
Certifications: Meets ASTM specification E982, Type IV requirements.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms