Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Bottles, Wide Neck, Amber, with Screw Cap
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU10861-838EA 0 INR InStock
VWRU10861-828 VWRU10861-838 VWRU89043-250 VWRU10861-784 VWRU10861-782 VWRU10861-844 VWRU10861-842 VWRU10861-786 VWRU10861-826 VWRU10861-958 VWRU10861-956 VWRU10861-846 VWRU10861-912
VWR®, Bottles, Wide Neck, Amber, with Screw Cap
Bottles Glass Bottles
Soda-lime glass.
  • Ideal for storage of solids
  • Wide-mouth design for efficient addition and removal of contents
Designed to protect contents from UV rays and ideal for light-sensitive products.
Certifications: Conforms to USP Type III and ASTM E438, Type II requirements.
Delivery information: Supplied as convenience packs with pulp/vinyl lined caps attached.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms