Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Autoclave deodorant
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
80080-888 80080-896
VWRU80080-888EA 0 INR InStock
VWRU80080-888 VWRU80080-896
VWR®, Autoclave deodorant
Deodorants and Air Fresheners
These steriliser deodorisers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture will dissolve the capsule and release a clean fresh scent.
  • Easy to use
  • Different fragrances available
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms