VWR®, Cryoboxes, PP
Supplier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
VWRU89084-934EA 942 INR
VWRU89084-934
VWR®, Cryoboxes, PP
Boxes Cryoboxes
PP, coloured.
  • Durable and autoclavable
  • An alphanumeric grid on the lid aids in sample identification
These boxes maintain their integrity under harsh freezer conditions and will not absorb water or degrade like cardboard storage boxes. A three-point hinge and positive catch provide quick access to samples and keep them secure when box is closed. Boxes are available in six colours; black is ideal for storage of light-sensitive samples.