Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Soft Nitrile, Nitrile Gloves
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU89038-270EA 0 INR InStock
VWRU89038-270 VWRU89038-267 VWRU89038-274 VWRU89038-272
VWR® Soft Nitrile, Nitrile Gloves
Gloves
Ideal for all industrial and laboratory tasks, these powder-free gloves are made of premium compounded nitrile for tactile sensitivity and comfort. Gloves protect against biohazards and a broad range of chemicals.
  • Non sterile, powder-free
  • 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
  • Made from premium nitrile for high quality and consistency
  • Gloves are fully textured and feature beaded cuffs
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms