Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Tool kit, deluxe, VWR®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWRU62510-100EA 0 INR InStock
VWRU62510-100
Tool kit, deluxe, VWR®
Deluxe tool kit includes 14 of the most essential tools for servicing laboratory equipment. Organized in heavy, blue vinyl, roll-up pouch with convenient see-through vinyl compartments. Two metal grommets provided for wall hanging. Tool handles cushioned in heavy rubber. Kit contains adjustable wrench, pocket oiler, contact tool, hex key set, pocket pen light, 3 pliers: pump, slip-joint, long nose, 6 screwdrivers: 1,6 mm tip jewelers, Phillips® with number 1 tip, offset and various-sized regulars (¹/₈, ³/₁₆ and ¹/₄" tip).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms