Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® TraceClean®, EPA vials
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR® TraceClean®
VWRU89094-202EA 0 INR InStock
VWRU89094-202
VWR® TraceClean®, EPA vials
Vials
These vials made from borosilicate glass, clear, type I are processed according to strict cleaning protocol, are lot traceable and include a Certificate of Analysis and specification form in each case.
  • TOC free
  • Ideal for pharmaceutical applications
Ordering information: Each vial is supplied with a PP cap with ultrasonically bonded fluoropolymer resin liner and a dust cover.
Packaging: All cases are custody-sealed to ensure content integrity.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms