Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sodium peroxodisulphate ≥95%, BAKER, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

V035-07EA 8291.18 INR InStock
V035-07 V035-01
Sodium peroxodisulphate ≥95%, BAKER, J.T.Baker®
Sodium peroxodisulphate
BAKER - A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).


Formula: Na₂S₂O₈
MW: 238.11 g/mol
Melting Pt: 100
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003501
CAS Number: 7775-27-1
EINECS: 231-892-1
UN: 1505
ADR: 5.1 III
Merck Index: 13,08729
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use = 95 %
Assay (Na₂S₂O₈) ≥95 %

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms