Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR®, Wipes, Pre-Saturated

Supplier: VWR BRAND PRIVATE LABEL USA

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

21910-110 21910-111
VWRU21910-110EA 0 INR InStock
VWRU21910-110 VWRU21910-111
VWR®, Wipes, Pre-Saturated
Wipes Disposable Wipes and Towels Presaturated Wipes
These non woven cellulose/polyester wipes are available premoistened with 70% reagent-grade isopropyl alcohol and 30% reagent-grade deionized water or with 100% pure reagent-grade deionized water. They are designed for environmentally sensitive areas.

  • Soft, lint-free, and will not scratch surfaces
  • Individually dispensed in a handy polyethylene canister
  • Unique opening allows easy removal of wipes
  • Snap-on sealed top helps eliminate contamination

Ideal for wiping optical parts, delicate glassware, electrodes, cuvettes, microscopes, and lenses.

Packaging: Wipes are supplied in a shrink-wrapped polyethylene canister, with 100 wipes per canister.

Caution: Do not use wipes on skin.

*The product(s) on this page are outside the scope of the EU Directive 2019/904

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms