Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Vortex mixers, MaxiMix®
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
BARNM16710-33QEA 0 INR InStock
BARNM16710-33Q
Vortex mixers, MaxiMix®
This mixer ensures fast, uniform mixing in continuous operation or touch-on mode.
  • Excellent for single and multiple tubes and small flasks with closed tops
  • Durable, white plastic case resists acids and alkalis
  • Compact design with 102 mm diameter foam rubber top
  • Push-button top-mounted ON/OFF switch
  • Cast aluminum base with suction cup rubber feet provides maximum stability
Delivery information: Includes one extra foam top, three-wire power cord and plug.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms