Puromycin dihydrochloride 10 mg/ml in water

Supplier: MIRUS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

MIRUMIR5940EA 42780 INR
MIRUMIR5940
Puromycin dihydrochloride 10 mg/ml in water
Puromycin dihydrochloride
Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544,44 g/mol
Storage Temperature: -20°C
MDL Number: MFCD00012691
CAS Number: 58-58-2
EINECS: 200-387-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR