Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Electroporation kits, Ingenio®
Supplier: MIRUS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
MIR50113 MIR50116 MIR50119 MIR50115 MIR50112 MIR50118
MIRUMIR50113EA 0 INR InStock
MIRUMIR50113 MIRUMIR50116 MIRUMIR50119 MIRUMIR50115 MIRUMIR50112 MIRUMIR50118
Electroporation kits, Ingenio®
Electroporation Systems Electroporation Kits
Ingenio® Electroporation Kits and Solution provide a universal, high efficiency, low toxicity solution for electroporation of DNA or siRNA into hard to transfect cell types.
  • Increased gene expression in several different cell types with minimal toxicity
  • Compatible with multiple conventional electroporation instruments, including Lonza Amaxa® Nucleofector®, Bio-Rad® Gene Pulser and Harvard-BTX® electroporators
  • Can be used for electroporation with both exponential decay as well as square wave forms
Ingenio® is provided a standalone solution or as part of a kit with cuvettes (0,2 and 0,4 cm) and cell droppers. The 0,2 cm cuvettes are part of the Ingenio® Electroporation Kit compatible with the Lonza-Amaxa® Nucleofector® II/2b, and the 0,4 cm cuvettes are part of the kit compatible with other electroporators, including Bio-Rad® and Harvard-BTX®.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms