Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Standards for the determination of Hydrocarbon Oil Index (HOI)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
87498.005EA 50250 INR InStock
VWRC87498.005
Standards for the determination of Hydrocarbon Oil Index (HOI)
Standards designed for the analysis of water and ground samples.
Certifications: ISO 9377-2.
Delivery information: Delivered with Certificate of Analysis, reference and batch number of source materials.
Packaging: 1,0 and 10 ml glass ampoules.
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms