Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Kjeldahl distillation unit, UDK 129

Supplier: VELP Scientifica

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

701-0224EA 0 INR InStock
VELPF30200120
Kjeldahl distillation unit, UDK 129
The UDK 129 unit is the entry level model for accurate and precise nitrogen and protein determination according to the Kjeldahl Method (TKN). This unit is the ideal solution for basic needs with the same key components and benefits of the more advanced models. It runs automatically, after setting sodium hydroxide addition and distillation time. The high precision pumps ensure constant accurate dosing of reagents and the cooling water is automatically stopped during pauses.

  • Automatic NaOH addition
  • Delay time (Devarda alloy analysis) and selectable distillation time
  • Safety lever and sensors to protect the user

All the UDK series distillation units have an alkali resistant technopolymer housing and accept different kinds of test tubes: straight tubes (100, 250, 400 ml and 1 L) or Kjeldahl balloon (500 ml).

Certifications: Conformity with multiple standards protocols AOAC, ISO, EPA, DIN etc.

Accessories information: Optional accessories such as test tubes, test tube spacer, test tube connection and IQ/OQ/PQ manuals are available on request.

Delivery information: Each unit comes ready to use and is supplied with 250 ml test tube, 250 ml collecting flask and pincer for test tubes.

Order Now
 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms