Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Avantor® Hichrom Vydac® 150HC C18, HPLC Columns
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
HICH5174060EA 246000 INR InStock
HICH5174060 HICH5174065
Avantor® Hichrom Vydac® 150HC C18, HPLC Columns
Vydac® 150HC media, developed specifically for peptide purification, is highly effective for purifying many small to medium-sized peptides with greater loading capacity and improved productivity.
  • Robust, high capacity 150 Å silica bonded with C18 for reversed phase
Caution: Vydac is a registered trademark of W.R.Grace and Co.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms