Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Handle for Whirl-Pak® racks
Supplier: Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Whirl-Pak®
NASCB01309WAEA 0 INR InStock
NASCB01309WA
Handle for Whirl-Pak® racks
Racks Bag Racks
This universal rack handle fits all sizes of racks, making it easy to carry a full rack in one hand. Attaches and detaches easily and quickly, so it never gets in the way.
  • Lightweight, strong, and comfortable to hold
  • Designed to prevent rack from tipping while it’s carried
Certifications: ISO 9001, ISO 13485.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms