Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Phenolphthalein 1% in ethanol

Supplier: BDH (US PRODUCED)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

BDHB7344-1EA 0 INR InStock
BDHB7344-1
Phenolphthalein 1% in ethanol
Phenolphthalein
Made with denatured ethanol.


Formula: C₂₀H₁₄O₄
MW: 318,33 g/mol
Melting Pt: 258 to 262 °C
Density: 1,3 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3,II
REACH: 01-2119498295-24
Merck Index: 14,07243
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg
Order Now
Specification Test Results
Appearance/colour Clear liquid

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms