Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Boston Round Bottles, Amber, Narrow Mouth
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU89042-908EA 0 INR InStock
VWRU89042-908 VWRU89042-918 VWRU89042-906 VWRU89042-928 VWRU89042-926 VWRU89042-914 VWRU89042-912 VWRU89042-922 VWRU89042-920 VWRU89042-876 VWRU89042-788 VWRU89042-930 VWRU89042-786 VWRU89045-404
VWR®, Boston Round Bottles, Amber, Narrow Mouth
Bottles Glass Bottles
These amber bottles are constructed of Type III soda-lime glass.
  • Good chemical and physical properties
  • Stable
  • For use in applications without high thermal loads
The amber bottles are designed to protect contents from UV rays and are ideal for light-sensitive products.
Delivery information: Bulk packs consist of shrink-wrapped glassware bundles (60 to 432 per wrap) packaged in corrugated cases. Bottle caps are included in separate bags within the case. Bottles sold without caps are packaged in bulk packs.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms