Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Drosophila Refrigerated Incubators with Peltier Technology
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU89510-750
VWR®, Drosophila Refrigerated Incubators with Peltier Technology
Incubators Cooled Incubators
Using innovative Peltier cooling technology, these drosphila incubators offer a 78% reduction in energy consumption over traditional compressor based refrigerated incubators.
  • Robust design
  • Whisper-quiet operation
  • Over-temperature thermostat and alarm
  • Independent over-temperature thermostat and alarm
These incubators are equipped with LED lighting and day/night controls to create ideal conditions for fruit flies. The robust cabinet design includes a mechanical convection system to provide improved temperature uniformity, digital temperature controller with display, and over temperature limit control. The compressor-free design creates a whisper-quiet operation.
Certifications: TUV, CE, UL and CSA listed.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms