Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Interval Timer
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU23609-218EA 2528 INR InStock
VWRU23609-218
VWR®, Interval Timer
Timers Mechanical Timers
This interval timer is adjustable from 0 - 60 minutes in one-minute intervals.
  • Easy to read dial
  • Easily audible bell alerts users to cycle end
  • Setting pointer to desired interval winds the time automatically
  • Indicating pointer always shows exact time remaining on easy-to-read numerals
  • Chemical resistant casing
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms