Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Titramax™, Digital burettes
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR® Titramax™
VWRU82017-948EA 145936 INR InStock
VWRU82017-948
VWR® Titramax™, Digital burettes
Burettes
Provides accurate and reliable titration results with the convenience of a large digital display, eliminating tedious meniscus readings.
  • Safer and more accurate than glass
  • Air purge and safety shutdown
  • Recirculating valve system
  • 360° swivel neck
The burette attaches directly to the reagent bottle, eliminating the hazards of pouring reagents. Designed to hold 50 ml of reagent so refilling during a titration does not compromise accuracy. With the unit's unique recirculating valves, air is purged from the system without loss of reagents. The valves can also be rotated to the safety position to prevent accidental discharge when the burette is not in use.
Tight control over reagent flow is maintained by easy-to-turn wheels on the side of the unit. Multiple titrations can be stored in memory with a maximum of 500 values. Using the optional interface and software package, stored values can be downloaded to a PC.
Delivery information: Includes a rechargeable NiCad battery, AC adapter/charger, and adapters to fit reagent bottles with 33 mm, 38 mm, [ST] 24/40, and [ST] 29/42 neck sizes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms