Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® Spinbar® Pyrex®, Magnetic stirring bars, round, glass
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU58948-488EA 0 INR InStock
VWRU58948-422 VWRU58948-488
VWR® Spinbar® Pyrex®, Magnetic stirring bars, round, glass
Stirrers Magnetic Stirring Bars
These glass encapsulated Alnico magnet stirring bars are useful for high temperature applications up to 274 °C.
  • Completely smooth surface offers 'zero absorption'; easy to clean
  • Meets US Pharmacopoeia specifications for Type I Borosilicate glass
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms