Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Laboratory refrigerators, spark-free, LR and LSR series, Lec
Supplier: Glen Dimplex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
471-0580EA 0 INR InStock
471-0580 471-0581
Laboratory refrigerators, spark-free, LR and LSR series, Lec
Refrigerators
Lec laboratory spark-free refrigerators offer safe, reliable storage of volatile liquids, reagents, poisons and samples. They have spark-free interiors to reduce the risk of internal explosions, CFC- and HCFC-free refrigeration systems ensuring minimal risk to the environment as well as locks to prevent theft or unauthorised access.
  • Environmentally friendly: CFC- and HFC-free
  • Temperature alarm
  • Low energy consumption
  • No internal fan
  • Doors with reversible hinges
Temperature: +2 to +10 Â°C
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms