Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

D-(+)-Glucose

Supplier: APPLICHEM LIFESCIENCE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Corn sugarD-GlucoseD-(+)-DextroseGlucolinᴅ-(+)-Glucose(2R,3S,4R,5R)-2,3,4,5,6-PentahydroxyhexanalGlucoseDextrose

A1422.5000EA 0 INR InStock
A1422.5000 A1422.1000
D-(+)-Glucose
D(+)-Glucose


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
UN: 0000
Merck Index: 14,04459
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/21/22/2302122.jpg
Order Now

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms