Iron(III) chloride 27% in aqueous solution, TECHNICAL

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ferric chlorideIron trichloride

24205.363EA 17348 INR
24205.363 24205.294
Iron(III) chloride 27% in aqueous solution, TECHNICAL
Iron (III) chloride
Formula: FeCl₃
MW: 162,21 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 260 °C
Density: 1,26 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8,III
Merck Index: 13,04048

Order Now

Specification Test Results

Assay 27 - 29 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR