EDTA disodium salt dihydrate
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethylenediaminetetraacetic acid disodium dihydrate
, Ethylenediaminetetra-acetic acid disodium salt dihydrate
, Edetate disodium dihydrate
, Ethylenediaminetetraacetic acid, disodium salt, dihydrate
, Ethylenediaminetetraacetic acid disodium salt dihydrate
, Kestranal® 2S 2 H2O
, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate
, Titriplex III
, Kestranal® 2S dihydrate
, Edetic acid disodium salt dihydrate
, Disodium dihydrogen ethylenediaminetetraacetate dihydrate
, Sequestrene Na₂
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 EINECS: 205-358-3 UN: 0000 Merck Index: 13,03543 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...