Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sodium perchlorate monohydrate, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: Perchloric acid sodium salt

2815.0100EA 8625 INR InStock
2815.0100
Sodium perchlorate monohydrate, J.T.Baker®
Sodium perchlorate monohydrate


Formula: NaClO₄·H₂O
MW: 140.46 g/mol
Melting Pt: 130 °C
Density: 2,02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149164
CAS Number: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 12,08798
Order Now

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms