Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Glutamine ≥98.5%, high purity

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: QL-GlutamineLevoglutamide(S)-(+)-GlutamineGlnH-Gln-OH

0374-500GEA 10753 INR InStock
0374-500G
L(+)-Glutamine ≥98.5%, high purity
L(+)-Glutamine
Non-animal origin. Fine white powder


Formula: C₅H₁₀N₂O₃
MW: 146,15 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 185…186 °C
Density: 1,5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
UN: 0000
Merck Index: 13,04484
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/32/00/16113200.jpg
Order Now
Specification Test Results
Arsenic <0.0001%
Ash ≤0.1%
Chloride <0.02%
Heavy Metals (as Pb) <0.001%
Iron ≤0.002%
Loss on Drying <0.2%
Purity > 98.5%
Solubility (10%, 2N HCl) pass
Specific Rotation (1 g/25 ml Water) 6.3 - 7.3°
Sulphate <0.03%
Thin Layer Chromatography (TLC) One Spot

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms