Urea
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Carbonyl diamide
, Carbamide
, Carbonyldiamide
, Diaminomethanone
A1360.9010EA
0
INR
A1360.9010
Urea
Urea
Formula:
NH₂CONH₂ MW: 60.06 g/mol Boiling Pt: 197 °C (1013 hPa) Melting Pt: 132,7 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 UN: 0000 REACH: 01-2119463277-33 Merck Index: 13,09935 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...