EDTA (ethylenediamine tetraacetic acid)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Ethylenediaminetetraacetic acid
, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid
, OmniPur® EDTA
, (Ethylenedinitrilo)tetraacetic acid
, Idranal® II
, Kestranal® A
, Edetic acid
, Ethylenedinitrilotetra-acetic acid
A5097.0500EA
0
INR
A5097.0500
EDTA (ethylenediamine tetraacetic acid)
EDTA (Ethylenediamine tetraacetic acid)
Formula:
C₁₀H₁₆N₂O₈ MW: 292.25 g/mol Boiling Pt: 614 °C (1013 hPa) Melting Pt: 220 °C Density: 0,86 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 UN: 0000 REACH: 01-2119486399-18 Merck Index: 13,03546 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...