Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Thiourea ≥99%, Proteomics Grade

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning
Synonyms: ThiocarbamideSulphourea

M226-100GEA 6223 INR InStock
M226-100G
Thiourea ≥99%, Proteomics Grade
Thiourea


Formula: NH₂CSNH₂
MW: 76,12 g/mol
Melting Pt: 174…177 °C
Density: 1,435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 13,09443
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/72/67/2297267.jpg
Order Now
Specification Test Results
Melting range 165 − 178 °C
Protease none detected
Purity ≥99%
Solubility (1%, Water) Pass
Appearance White powder

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms