Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ethylenediamine ≥99%, TECHNICAL

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger
Synonyms: 1,2-Diaminoethane1,2-Ethanediamine

24039.290EA 5144 INR InStock
24039.290
Ethylenediamine ≥99%, TECHNICAL
Ethylenediamine


Formula: C₂H₈N₂
MW: 60,1 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 8,5 °C
Density: 0,898 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8 (3),II
Merck Index: 13,03829
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/99/69/2299969.jpg
Order Now
Specification Test Results
Assay Min. 99 %

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms