5-Sulphosalicylic acid dihydrate 99-101%, white crystals ACS

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Hydroxy-5-sulphobenzoic acid dihydrate

0610-250GEA 8223 INR
0610-250G
5-Sulphosalicylic acid dihydrate 99-101%, white crystals ACS
5-Sulphosalicylic acid dihydrate
Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254,22 g/mol
Melting Pt: 224 °C
Density: 1,695 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8,III
Merck Index: 13,09054
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/90/04/2299004.jpg

Order Now

Specification Test Results

Ash <0.1%
Chloride ≤0.001%
Heavy metals ≤0.002%
Insolubles ≤0.02%
Iron ≤0.001%
Purity 99 – 101%
Salicylic acid ≤0.04%
Sulphate ≤0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR