Fluorescein, TECHNICAL
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
6539.1000EA
20002
INR
6539.1000
Fluorescein, TECHNICAL
Fluorescein
Formula:
C₂₀H₁₂O₅ MW: 332,31 g/mol Melting Pt: 314…316 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005050 CAS Number: 2321-07-5 EINECS: 219-031-8 UN: 0000 Merck Index: 13,04185 |
Specification Test Results
Ignition residue (SO4) | Max. 2 % |
Loss on drying (100-105°C) | Max. 1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...