Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Dimethyl sulphoxide ≥99.9%, BioChemica®, sterile filtered

Supplier: APPLICHEM LIFESCIENCE

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: DMSOMethyl sulphoxideMethanesulphinylmethaneDimethyl(oxido)sulphur

A7248.0010EA 0 INR InStock
A7248.0010
Dimethyl sulphoxide ≥99.9%, BioChemica®, sterile filtered
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78,14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18,5 °C
Density: 1,101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
UN: 0000
Merck Index: 13,03285
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
Endotoxins max. 40 E.U./g
Assay (GC) min. 99.9 %
Appearance clear, colorless
Water (K.F.) max. 0.5 %
A 275 nm (1 cm/water HPLC grade) max. 0.30

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms