Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Sterile PETG Single-Use Bottles with Aseptic Connector
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWRI734-3216EA 96948 INR InStock
VWRI734-3216
VWR®, Sterile PETG Single-Use Bottles with Aseptic Connector
Sterile PETG containers provide the maximum amount of flexibility and reliability.
  • Platinum cured silicone tubing
  • CPC® AseptiQuik® S Connector
  • USP Class VI
  • Animal-free or processed in accordance to EMEA/410/01
  • Assemblies are manufactured in certified ISO Class 7 cleanroom
  • Gamma irradiated to Sterility Assurance Leve (SAL) 10⁻⁶
  • Each assembly comes individually double-bagged
  • Multiple bottle sizes allow for process-specific flexibility
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and applications. Using assemblies with an aseptic connector eliminates the need for a laminar hood as well as additional equipment (ie. welder), lessens the risk of contamination, and saves valuable time. The AseptiQuik® “Flip-Click-Pull” design enables users to quickly and easily transfer small volumes of media. When used with sterile aseptic connector tube sets, custom possibilities are endless. Other aseptic connectors are available upon request. Please consult a Avantor Account Manager to discuss custom single-use needs.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms