Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

5907-01EA 14133.43 INR InStock
5907-01
Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®
Barium chloride


Formula: BaCl₂
MW: 208,24 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: ~ 963 °C
Density: 1,00 g/cm³
Storage Temperature: ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (BaCl₂·2H₂O), w/v 11.6 - 12.4 %
Meets test solution (T.S.) specifications for testing USP/NF monographs.

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms