Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Zinc, AAS standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
6474-04EA 34415 INR InStock
BAKR6474-04
Zinc, AAS standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
Prepared by dissolution of zinc metal of 99.99% spectral purity. The metal content of the solution is confirmed by tritation with EDTA, traceable to NIST SRM 915.
  • Standardised with NIST standard chemicals
Caution: 10,000 µg/ml= (1,00% w/v)= 10000 mg/L
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms