Caffeine, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 7-Methyltheobromine1,3,7-Trimethylxanthine2,6-Dihydroxy-1,3,7-trimethylpurineMethyltheobromine1,3,7-Trimethyl-2,6-dioxopurine3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione

Management of Change (MOC) category = N

E268-07EA 25010.57 INR
E268-07
Caffeine, J.T.Baker®
Caffeine
Formula: C₈H₁₀N₄O₂
MW: 194,19 g/mol
Melting Pt: 233,..237 °C
Density: 1,23 g/cm³
Storage Temperature: ambient
MDL Number: MFCD00005758
CAS Number: 58-08-2
EINECS: 200-362-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,01636
https://b2b-in-gqp.vwr.com/stibo/web/std.lang.all/79/46/2297946.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Melting Point 233 - 237 °C
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR