Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Calcium carbonate 99.95-100.05%, powder, AR® ACS chelometric standard, Macron Fine Chemicals™

Supplier: MACRON (A AVANTOR BRAND)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:


4071-12EA 12848.57 INR InStock
4071-12 4071-02 MACR4071-12
Calcium carbonate 99.95-100.05%, powder, AR® ACS chelometric standard, Macron Fine Chemicals™
Calcium carbonate


Formula: CaCO₃
MW: 100,09 g/mol
Melting Pt: 825 °C
Density: 2,71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
UN: 0000
Merck Index: 13,01658
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CaCO₃) 99.95 - 100.05 %
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Appearance (fine, white powder) Passes Test
ACS - Heavy Metals (as Pb) ≤ 0.001 %
ACS - Alkalinity Passes Test
ACS - Ammonium (NH₄) ≤ 0.003 %
ACS - Insoluble in Dilute HCl ≤ 0.01 %
ACS - Insoluble in HCl ≤ 0.005 %
ACS - Oxidizing Substances (as NO₃) ≤ 0.005 %
ACS - Silica (SiO₂) ≤ 0.001 %
ACS - Sulfate (SO₄) ≤ 0.005 %
ACS - Fluoride (F) ≤ 0.0015 %
ACS - Barium (Ba) ≤ 0.005 %
ACS - Iron (Fe) ≤ 0.002 %
ACS - Magnesium (Mg) ≤ 0.01 %
ACS - Potassium (K) ≤ 0.01 %
ACS - Sodium (Na) ≤ 0.01 %
ACS - Silicon (Si), For Information Only
ACS - Strontium (Sr) ≤ 0.1 %

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms