Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Diatomaceous earth, BAKER ANALYZED® 48 µm Pest_Residual, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

3371-01EA 23469.5 INR InStock
3371-01 3371-05
Diatomaceous earth, BAKER ANALYZED® 48 µm Pest_Residual, J.T.Baker®
Diatomaceous earth
For use in pesticide residue analysis after petroleum ether extraction.


Formula: SiO₂
MW: 60.08 g/mol
Density: 2.30 g/cm³
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Loss on Drying at 110°C ≤ 0.2 %
pH of 5% Slurry at 25°C 8.0 - 10.0
Petroleum Ether Extractable Residue, For Information Only

 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms