Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Benchtop autoclaves, Elara Class B
Supplier: Tuttnauer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
TUTTELARA11-DEA 0 INR InStock
TUTTELARA11-D
Benchtop autoclaves, Elara Class B
Sterilisers Autoclaves (Steam Sterilisers)
These models are reliable and robust Class B autoclaves designed for the needs of modern clinics where autoclaves are expected to meet sophisticated international standards and operate non-stop every day. Suitable for the sterilisation processing needs of medical, dental and veterinary clinics.
  • Self-locking door designed to easily close/open door with one hand
  • Bright LCD panel for easy reading, with multilingual interface (26 languages)
  • Automatic safety shutoff prevents overheating
  • Maintenance free with reduced running cost
  • Built-in system for checking water quality
Durable 316L type stainless steel chamber and electro-polish finish door.
Class B performance is achieved with a built-in vacuum pump used for fractionated pre-vacuum air removal eliminating air pockets from
all load types and maximising efficient steam penetration throughout the entire load. After the sterilisation stage the vacuum pump is used for post-vacuum drying.
Features a user-friendly three button keypad enables the user to easily browse through the sterilisation cycle program settings and user directories.
Certifications: Complies with ISO 9001:2008, EN ISO 13485, AAMI/ANSI ST-55:2010,EN 13060, EN 61010-1, EN 61326-1, EN 61010-2-40, EN 17665-1.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms