Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
pMCS minP-Tluc16 vector for luciferase assays, Pierce™
Supplier: Thermo Fisher Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PIER88236EA 0 INR InStock
PIER88236
pMCS minP-Tluc16 vector for luciferase assays, Pierce™
The pMCS minP-Tluc16 vector is a transcriptional reporter vector that encodes the smallest known luciferase, TurboLuc™16 (Tluc16) luciferase, as the reporter.
  • Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systems
  • Optimised minimal core promoter (minP) and 5′ UTR region for efficient expression of Tluc16 luciferase
  • Multiple cloning sites provide versatility for transfer of regulatory elements from one plasmid to another
The vector contains multiple cloning sites (MCS) and a core minimal promoter (minP) upstream of the reporter gene for the insertion of regulatory elements lacking promoter activity.
Tluc16 luciferase is a 16 kDa, novel, intracellular luciferase derived from the marine copedod Metridia luciferase family. The wild-type luciferase has been modified to reduce its size, increase its brightness, and enable its intracellular expression.
A synthetic polyA terminator and a Transcriptional Pause Site (TPS) are included upstream of the MCS to minimize non-specific transcriptional read-through.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms