Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Cryovials, 2D Barcoded
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU75852-324EA 46285 INR InStock
VWRU75852-326 VWRU75852-324
VWR®, Cryovials, 2D Barcoded
Vials
These 2 ml cryovials feature unique 2D barcodes and human readable characters.
  • Designed to read from the base through opening for each tube
  • Superior quality readability even in harsh conditions due to laser etching technology
  • Sterilised 10⁻⁶. Certified Dnase-, Rnase-, Pyrogen-, and DNA-free
Cryovials are designed for storing biological material, human, or animal cells at a temperature as low as –196 °C. The black 2D insert at the bottom of the vial can be read by standard 2D barcode readers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms