Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Beard cover, irradiated
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
VWR®
VWRU414004-506EA 14172 INR InStock
VWRU414004-506
VWR®, Beard cover, irradiated
Disposable Garments
The beard cover is manufactured from strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in nonhazardous environments.
  • Lowest level of particle shedding, non-linting
  • First line barrier protection
  • Manufactured and packaged in an ISO Class 8 cleanroom
Gamma irradiated to Sterility Assurance Level of 10-4.
Weight: 16,82 g
Certifications: Tested and manufactured in an ISO certified facility, lot-controlled and validated through independent laboratory testing.
Packaging: Packed in sealed polyethylene bags, 10 per bag, contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms