Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
BeneSphera™, Chikungunya IgG/ IgM Rapid Test
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
15001386 15001387
BENP15001386EA 8750 INR InStock
BENP15001386 BENP15001387
BeneSphera™, Chikungunya IgG/ IgM Rapid Test
Clinical Diagnostic Supplies Clinical Reagents and Kits
The BeneSphera™ Chikungunya IgG/IgM Rapid Test qualitatively detects IgG and IgM antibodies to Chikungunya in human whole blood, serum or plasma samples. This test applies lateral flow immunochromatography and is intended to be used as a screening test and as an aid in the diagnosis of infection with CHIK.
  • Detection of IgM antibodies against all isolates
  • No cross reactivity with other mosquito borne bacterial/viral infections has been reported
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms