Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Acetone FCC, Multi-Compendial, Macron Fine Chemicals™

Supplier: MACRON AVANTOR BRAND

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Danger

MACR4952-06EA 0 INR InStock
MACR4952-06 MACR4952-08
Acetone FCC, Multi-Compendial, Macron Fine Chemicals™
Acetone


Formula: CH₃COCH₃
Boiling Pt: 56,2 °C (1013 hPa)
Melting Pt: –95,4 °C
Density: 0,792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=e42f85852579489db8cf68a147710248276677d6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Food GMP Manufactured Product
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets F.C.C. Requirements
Meets J.P. Chemical Specifications
Store in airtight container.
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
FCC - Assay (C₆H₉N₃O₂·HCl) (dried basis) 98.5 - 101.5 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤ 5 mg/kg
FCC - Loss on Drying at 105°C ≤ 0.3 %
FCC - Residue after Ignition ≤ 0.10 %
FCC - Specific Rotation [Å]²º^D (+) 8.5 - 10.5 Degree
EP/BP - Assay (C₆H₉N₃O₂·HCl) (dried basis) 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification F Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 3.0 - 5.0
EP/BP - Specific Rotation [Å]²º^D (+) 9.2 - 10.6 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying at 150°C 7.0 - 10.0 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Identification Passes Test
JP - Optical Rotation (+) 9.2 - 10.6
JP - pH 3.5 - 4.5
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Iron (Fe) ≤ 10 ppm
JP - Related Substances Passes Test
JP - Water 7.2 - 10.0 %
JP - Residue on Ignition ≤ 0.10 %
JP - Assay (anhydrous basis) 99.0 - 101.0 %
Microbial Testing – Total Aerobic Microbial Count (USP) ≤ 10 cfu/g
Microbial Testing – Total Yeast and Mold Count (USP) ≤ 10 cfu/g
Endotoxin Concentration (EU/g) ≤ 10
Trace Impurities - Cadmium (Cd) ≤ 200 ppb
Trace Impurities - Lead (Pb) ≤ 500 ppb
Trace Impurities - Arsenic (As) ≤ 1500 ppb
Trace Impurities - Mercury (Hg) ≤ 300 ppb
Trace Impurities - Cobalt (Co) ≤ 500 ppb
Trace Impurities - Vanadium (V) ≤ 1000 ppb
Trace Impurities - Nickel (Ni) ≤ 2000 ppb
Trace Impurities - Thallium (Tl) ≤ 800 ppb
Trace Impurities - Gold (Au) ≤ 10000 ppb
Trace Impurities - Palladium (Pd) ≤ 1000 ppb
Trace Impurities - Iridium (Ir) ≤ 1000 ppb
Trace Impurities - Osmium (Os) ≤ 1000 ppb
Trace Impurities - Rhodium (Rh) ≤ 1000 ppb
Trace Impurities - Ruthenium (Ru) ≤ 1000 ppb
Trace Impurities - Selenium (Se) ≤ 8000 ppb
Trace Impurities - Silver (Ag) ≤ 1000 ppb
Trace Impurities - Platinum (Pt) ≤ 1000 ppb
Trace Impurities - Lithium (Li) ≤ 25000 ppb
Trace Impurities - Antimony (Sb) ≤ 9000 ppb
Trace Impurities - Barium (Ba) ≤ 70000 ppb
Trace Impurities - Molybdenum (Mo) ≤ 150000 ppb
Trace Impurities - Copper (Cu) ≤ 30000 ppb
Trace Impurities - Tin (Sn) ≤ 60000 ppb
Trace Impurities - Chromium (Cr) ≤ 110000 ppb
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP <232>, EP 5.20) - Information on
elemental impurities for this product is available on the associated
Product Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms