Nucleosil Silica, HPLC Columns
Supplier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Nucleosil
NC100510C5SKEA 0 INR
NC100510C5SK NC100-3-150AS NC100-7-10C5 NC100-5-50AM NC100-3-250AF NC100-5-200A NC100-5-10C5 NC100-5-250AM NC100-5-150A
Nucleosil Silica, HPLC Columns
Chromatography Columns
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity whilst the 120 Å exhibits a lower backpressure.
  • Good mechanical stability
  • Good batch to batch reproducibility