Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
HPLC columns, Develosil®, ODS-HG
Supplier: Hichrom Limited
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
HG-5-250AEA 0 INR InStock
HICHHG-5-150A HICHHG-5-250A HICHHG-3-150A
HPLC columns, Develosil®, ODS-HG
Develosil® ODS-HG is the most stable column under acidic conditions (pH 1 to 9) and can be used with up to 0,5% TFA.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms